Learn More
ABclonal Technology MAN2A1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A102271000UL
Encompass_Preferred
This gene encodes a glycosyl hydrolase that localizes to the Golgi and catalyzes the final hydrolytic step in the asparagine-linked oligosaccharide (N-glycan) maturation pathway. Mutations in the mouse homolog of this gene have been shown to cause a systemic autoimmune disease similar to human systemic lupus erythematosus.
Catalog No. 50-001-35875