Learn More
ABclonal Technology SPG7 Rabbit pAb
SDP
Supplier:  ABclonal Technology A102491000UL
Encompass_Preferred
This gene encodes a mitochondrial metalloprotease protein that is a member of the AAA family. Members of this protein family share an ATPase domain and have roles in diverse cellular processes including membrane trafficking, intracellular motility, organelle biogenesis, protein folding, and proteolysis. Mutations in this gene cause autosomal recessive spastic paraplegia 7. Two transcript variants encoding distinct isoforms have been identified.
Catalog No. 50-001-35221