Learn More
ABclonal Technology NR2F2 Rabbit pAb
SDP
Catalog No. 5000135284
Encompass_Preferred
This gene encodes a member of the steroid thyroid hormone superfamily of nuclear receptors. The encoded protein is a ligand inducible transcription factor that is involved in the regulation of many different genes. Alternate splicing results in multiple transcript variants.
Catalog No. 50-001-35284 Supplier ABclonal Technology Supplier No. A102511000UL
This gene encodes a member of the steroid thyroid hormone superfamily of nuclear receptors. The encoded protein is a ligand inducible transcription factor that is involved in the regulation of many different genes. Alternate splicing results in multiple transcript variants.