Learn More
ABclonal Technology NR2F2 Rabbit pAb
SDP
Supplier:  ABclonal Technology A10251500UL
Encompass_Preferred
This gene encodes a member of the steroid thyroid hormone superfamily of nuclear receptors. The encoded protein is a ligand inducible transcription factor that is involved in the regulation of many different genes. Alternate splicing results in multiple transcript variants.
Catalog No. 50-001-36740