Learn More
ABclonal Technology XPNPEP2 Rabbit pAb
SDP
Supplier:  ABclonal Technology A10255500UL
Encompass_Preferred
Aminopeptidase P is a hydrolase specific for N-terminal imido bonds, which are common to several collagen degradation products, neuropeptides, vasoactive peptides, and cytokines. Structurally, the enzyme is a member of the pita bread fold family and occurs in mammalian tissues in both soluble and GPI-anchored membrane-bound forms. A membrane-bound and soluble form of this enzyme have been identified as products of two separate genes.
Catalog No. 50-001-37671