Learn More
ABclonal Technology 14-3-3 sigma Rabbit pAb
SDP
Supplier:  ABclonal Technology A10261000UL
Encompass_Preferred
This gene encodes a cell cycle checkpoint protein. The encoded protein binds to translation and initiation factors and functions as a regulator of mitotic translation. In response to DNA damage this protein plays a role in preventing DNA errors during mitosis.
Catalog No. 50-001-37752