Learn More
ABclonal Technology USP13 Rabbit pAb
SDP
Catalog No. 5000138648
Encompass_Preferred
Enables several functions, including BAT3 complex binding activity, chaperone binding activity, and cysteine-type peptidase activity. Involved in several processes, including maintenance of unfolded protein involved in ERAD pathway, regulation of cellular catabolic process, and regulation of transcription, DNA-templated. Acts upstream of or within protein deubiquitination and protein stabilization. Predicted to be located in nucleoplasm. Predicted to be active in cytosol and nucleus.
Catalog No. 50-001-38648 Supplier ABclonal Technology Supplier No. A10264500UL
Enables several functions, including BAT3 complex binding activity, chaperone binding activity, and cysteine-type peptidase activity. Involved in several processes, including maintenance of unfolded protein involved in ERAD pathway, regulation of cellular catabolic process, and regulation of transcription, DNA-templated. Acts upstream of or within protein deubiquitination and protein stabilization. Predicted to be located in nucleoplasm. Predicted to be active in cytosol and nucleus.