Learn More
ABclonal Technology SLC4A7 Rabbit pAb
SDP
Supplier:  ABclonal Technology A102711000UL
Encompass_Preferred
This locus encodes a sodium bicarbonate cotransporter. The encoded transmembrane protein appears to transport sodium and bicarbonate ions in a 11 ratio, and is thus considered an electroneutral cotransporter. The encoded protein likely plays a critical role in regulation of intracellular pH involved in visual and auditory sensory transmission. Alternatively spliced transcript variants encoding distinct isoforms have been described.
Catalog No. 50-001-37674