Learn More
ABclonal Technology SCN3B Rabbit pAb
SDP
Supplier:  ABclonal Technology A103161000UL
Encompass_Preferred
Voltage-gated sodium channels are transmembrane glycoprotein complexes composed of a large alpha subunit and one or more regulatory beta subunits. They are responsible for the generation and propagation of action potentials in neurons and muscle. This gene encodes one member of the sodium channel beta subunit gene family, and influences the inactivation kinetics of the sodium channel. Two alternatively spliced variants, encoding the same protein, have been identified.
Catalog No. 50-001-37709