Learn More
ABclonal Technology CISD1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A103171000UL
Encompass_Preferred
This gene encodes a protein with a CDGSH iron-sulfur domain and has been shown to bind a redox-active [2Fe-2S] cluster. The encoded protein has been localized to the outer membrane of mitochondria and is thought to play a role in regulation of oxidation. Genes encoding similar proteins are located on chromosomes 4 and 17, and a pseudogene of this gene is located on chromosome 2.
Catalog No. 50-001-37710