Learn More
ABclonal Technology SLC28A3 Rabbit pAb
SDP
Supplier:  ABclonal Technology A103201000UL
Encompass_Preferred
Nucleoside transporters, such as SLC28A3, regulate multiple cellular processes, including neurotransmission, vascular tone, adenosine concentration in the vicinity of cell surface receptors, and transport and metabolism of nucleoside drugs. SLC28A3 shows broad specificity for pyrimidine and purine nucleosides (Ritzel et al., 2001 [PubMed 11032837]).
Catalog No. 50-001-35207