Learn More
ABclonal Technology LRRC4 Rabbit pAb
SDP
Catalog No. 5000137701
Encompass_Preferred
Predicted to be involved in modulation of chemical synaptic transmission and synapse organization. Predicted to act upstream of or within synapse organization. Predicted to be located in neuron spine and postsynaptic membrane. Predicted to be active in Schaffer collateral - CA1 synapse, glutamatergic synapse, and plasma membrane. Predicted to be integral component of postsynaptic density membrane.
Catalog No. 50-001-37701 Supplier ABclonal Technology Supplier No. A10321500UL
Predicted to be involved in modulation of chemical synaptic transmission and synapse organization. Predicted to act upstream of or within synapse organization. Predicted to be located in neuron spine and postsynaptic membrane. Predicted to be active in Schaffer collateral - CA1 synapse, glutamatergic synapse, and plasma membrane. Predicted to be integral component of postsynaptic density membrane.