Learn More
ABclonal Technology MS4A8B Rabbit pAb
SDP
Supplier:  ABclonal Technology A10330500UL
Encompass_Preferred
This gene encodes a member of the membrane-spanning 4A gene family. Members of this protein family are characterized by common structural features and similar intron/exon splice boundaries and display unique expression patterns among hematopoietic cells and nonlymphoid tissues. The gene encoding this protein is localized to 11q12.3, among a cluster of family members.
Catalog No. 50-001-38306