Learn More
ABclonal Technology L3MBTL2 Rabbit pAb
SDP
Catalog No. 5000137326
Encompass_Preferred
Enables methylated histone binding activity. Predicted to be involved in negative regulation of transcription, DNA-templated. Predicted to act upstream of or within several processes, including ectoderm development, regulation of histone modification, and stem cell proliferation. Located in nucleus.
Catalog No. 50-001-37326 Supplier ABclonal Technology Supplier No. A103311000UL
Add to Cart
Add to Cart
Enables methylated histone binding activity. Predicted to be involved in negative regulation of transcription, DNA-templated. Predicted to act upstream of or within several processes, including ectoderm development, regulation of histone modification, and stem cell proliferation. Located in nucleus.