Learn More
ABclonal Technology RAB34 Rabbit pAb
SDP
Catalog No. 5000138183
Encompass_Preferred
This gene encodes a protein belonging to the RAB family of proteins, which are small GTPases involved in protein transport. This family member is a Golgi-bound member of the secretory pathway that is involved in the repositioning of lysosomes and the activation of macropinocytosis. Alternative splicing of this gene results in multiple transcript variants. An alternatively spliced transcript variant produces the nine-amino acid residue-repeats (NARR) protein, which is a functionally distinct nucleolar protein resulting from a different reading frame.
Catalog No. 50-001-38183 Supplier ABclonal Technology Supplier No. A10332500UL
This gene encodes a protein belonging to the RAB family of proteins, which are small GTPases involved in protein transport. This family member is a Golgi-bound member of the secretory pathway that is involved in the repositioning of lysosomes and the activation of macropinocytosis. Alternative splicing of this gene results in multiple transcript variants. An alternatively spliced transcript variant produces the nine-amino acid residue-repeats (NARR) protein, which is a functionally distinct nucleolar protein resulting from a different reading frame.