Learn More
ABclonal Technology SNX18 Rabbit pAb
SDP
Supplier:  ABclonal Technology A103371000UL
Encompass_Preferred
This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. This protein does not contain a coiled coil region, like some family members, but contains a SH3 domain. Multiple transcript variants encoding different isoforms have been found for this gene.
Catalog No. 50-001-37723