Learn More
ABclonal Technology DIAPH3 Rabbit pAb
SDP
Supplier:  ABclonal Technology A10351500UL
Encompass_Preferred
This gene encodes a member of the diaphanous subfamily of the formin family. Members of this family are involved in actin remodeling and regulate cell movement and adhesion. Mutations in this gene are associated with autosomal dominant auditory neuropathy 1. Multiple transcript variants encoding different isoforms have been found for this gene.
Catalog No. 50-001-38204