Learn More
ABclonal Technology NELL1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A103581000UL
Encompass_Preferred
This gene encodes a cytoplasmic protein that contains epidermal growth factor (EGF)-like repeats. The encoded heterotrimeric protein may be involved in cell growth regulation and differentiation. A similar protein in rodents is involved in craniosynostosis. Alternative splicing results in multiple transcript variants.
Catalog No. 50-001-35429