Learn More
ABclonal Technology HNRPLL Rabbit pAb
SDP
Supplier:  ABclonal Technology A103601000UL
Encompass_Preferred
HNRNPLL is a master regulator of activation-induced alternative splicing in T cells. In particular, it alters splicing of CD45 (PTPRC, MIM 151460), a tyrosine phosphatase essential for T-cell development and activation (Oberdoerffer et al., 2008 [PubMed 18669861]).
Catalog No. 50-001-35431