Learn More
ABclonal Technology NOP14 Rabbit pAb
SDP
Supplier:  ABclonal Technology A103611000UL
Encompass_Preferred
This gene encodes a protein that plays a role in pre-18s rRNA processing and small ribosomal subunit assembly. The encoded protein may be involved in the regulation of pancreatic cancer cell proliferation and migration. Alternative splicing results in multiple transcript variants.
Catalog No. 50-001-35421