Learn More
ABclonal Technology AADAC Rabbit pAb
SDP
Catalog No. 5000135433
Encompass_Preferred
Microsomal arylacetamide deacetylase competes against the activity of cytosolic arylamine N-acetyltransferase, which catalyzes one of the initial biotransformation pathways for arylamine and heterocyclic amine carcinogens
Catalog No. 50-001-35433 Supplier ABclonal Technology Supplier No. A103651000UL
Microsomal arylacetamide deacetylase competes against the activity of cytosolic arylamine N-acetyltransferase, which catalyzes one of the initial biotransformation pathways for arylamine and heterocyclic amine carcinogens