Learn More
ABclonal Technology AADAC Rabbit pAb
SDP
Supplier:  ABclonal Technology A10365500UL
Encompass_Preferred
Microsomal arylacetamide deacetylase competes against the activity of cytosolic arylamine N-acetyltransferase, which catalyzes one of the initial biotransformation pathways for arylamine and heterocyclic amine carcinogens
Catalog No. 50-001-37516