Learn More
ABclonal Technology CNDP2 Rabbit pAb
SDP
Catalog No. 5000137514
Encompass_Preferred
CNDP2, also known as tissue carnosinase and peptidase A (EC 3.4.13.18), is a nonspecific dipeptidase rather than a selective carnosinase (Teufel et al., 2003 [PubMed 12473676]).
Catalog No. 50-001-37514 Supplier ABclonal Technology Supplier No. A10381500UL
CNDP2, also known as tissue carnosinase and peptidase A (EC 3.4.13.18), is a nonspecific dipeptidase rather than a selective carnosinase (Teufel et al., 2003 [PubMed 12473676]).