Learn More
ABclonal Technology ERAL1 Rabbit pAb
SDP
Catalog No. 5000138260
Encompass_Preferred
The protein encoded by this gene is a GTPase that localizes to the mitochondrion. The encoded protein binds to the 3 terminal stem loop of 12S mitochondrial rRNA and is required for proper assembly of the 28S small mitochondrial ribosomal subunit. Deletion of this gene has been shown to cause mitochondrial dysfunction, growth retardation, and apoptosis. Several transcript variants encoding different isoforms have been found for this gene.
Catalog No. 50-001-38260 Supplier ABclonal Technology Supplier No. A10382500UL
The protein encoded by this gene is a GTPase that localizes to the mitochondrion. The encoded protein binds to the 3 terminal stem loop of 12S mitochondrial rRNA and is required for proper assembly of the 28S small mitochondrial ribosomal subunit. Deletion of this gene has been shown to cause mitochondrial dysfunction, growth retardation, and apoptosis. Several transcript variants encoding different isoforms have been found for this gene.