Learn More
ABclonal Technology RAB3D Rabbit pAb
SDP
Supplier:  ABclonal Technology A103901000UL
Encompass_Preferred
Enables myosin V binding activity. Involved in bone resorption and positive regulation of regulated secretory pathway. Located in cytoplasmic microtubule and secretory vesicle.
Catalog No. 50-001-38262