Learn More
ABclonal Technology REEP5 Rabbit pAb
SDP
Catalog No. 5000138186
Encompass_Preferred
Predicted to be involved in endoplasmic reticulum organization and regulation of intracellular transport. Located in endoplasmic reticulum tubular network.
Catalog No. 50-001-38186 Supplier ABclonal Technology Supplier No. A10392500UL
Predicted to be involved in endoplasmic reticulum organization and regulation of intracellular transport. Located in endoplasmic reticulum tubular network.