Learn More
ABclonal Technology MIEF1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A103961000UL
Encompass_Preferred
Enables ADP binding activity, GDP binding activity, and identical protein binding activity. Involved in several processes, including positive regulation of mitochondrial fission, positive regulation of mitochondrial translation, and positive regulation of protein targeting to membrane. Located in mitochondrial matrix and mitochondrial outer membrane. Colocalizes with mitochondrial large ribosomal subunit.
Catalog No. 50-001-36430