Learn More
ABclonal Technology STOML2 Rabbit pAb
SDP
Catalog No. 5000137341
Encompass_Preferred
Enables GTPase binding activity and cardiolipin binding activity. Involved in several processes, including inorganic cation transmembrane transport, positive regulation of cardiolipin metabolic process, and positive regulation of mitochondrial DNA replication. Located in membrane raft, mitochondrial inner membrane, and mitochondrial intermembrane space. Is extrinsic component of plasma membrane. Colocalizes with several cellular components, including COP9 signalosome, T cell receptor complex, and immunological synapse.
Catalog No. 50-001-37341 Supplier ABclonal Technology Supplier No. A10398500UL
Enables GTPase binding activity and cardiolipin binding activity. Involved in several processes, including inorganic cation transmembrane transport, positive regulation of cardiolipin metabolic process, and positive regulation of mitochondrial DNA replication. Located in membrane raft, mitochondrial inner membrane, and mitochondrial intermembrane space. Is extrinsic component of plasma membrane. Colocalizes with several cellular components, including COP9 signalosome, T cell receptor complex, and immunological synapse.