Learn More
ABclonal Technology YME1L1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A10402500UL
Encompass_Preferred
The protein encoded by this gene is the human ortholog of yeast mitochondrial AAA metalloprotease, Yme1p. It is localized in the mitochondria and can functionally complement a yme1 disruptant yeast strain. It is proposed that this gene plays a role in mitochondrial protein metabolism and could be involved in mitochondrial pathologies. Three transcript variants encoding different isoforms have been found for this gene.
Catalog No. 50-001-38279