Learn More
ABclonal Technology GNA14 Rabbit pAb
SDP
Supplier:  ABclonal Technology A104051000UL
Encompass_Preferred
This gene encodes a member of the guanine nucleotide-binding, or G protein family. G proteins are heterotrimers consisting of alpha, beta and gamma subunits. The encoded protein is a member of the alpha family of G proteins, more specifically the alpha q subfamily of G proteins. The encoded protein may play a role in pertussis-toxin resistant activation of phospholipase C-beta and its downstream effectors.
Catalog No. 50-001-36429