Learn More
ABclonal Technology SEMA4F Rabbit pAb
SDP
Catalog No. 5000135270
Encompass_Preferred
This gene encodes a transmembrane class IV semaphorin family protein, which plays a role in neural development. This gene may be involved in neurogenesis in prostate cancer, the development of neurofibromas, and breast cancer tumorigenesis. Alternative splicing results in multiple transcript variants.
Catalog No. 50-001-35270 Supplier ABclonal Technology Supplier No. A10432500UL
This gene encodes a transmembrane class IV semaphorin family protein, which plays a role in neural development. This gene may be involved in neurogenesis in prostate cancer, the development of neurofibromas, and breast cancer tumorigenesis. Alternative splicing results in multiple transcript variants.