Learn More
ABclonal Technology CYC1 Rabbit pAb
SDP
Catalog No. 5000137644
Encompass_Preferred
This gene encodes a subunit of the cytochrome bc1 complex, which plays an important role in the mitochondrial respiratory chain by transferring electrons from the Rieske iron-sulfur protein to cytochrome c. Mutations in this gene may cause mitochondrial complex III deficiency, nuclear type 6.
Catalog No. 50-001-37644 Supplier ABclonal Technology Supplier No. A10449500UL
This gene encodes a subunit of the cytochrome bc1 complex, which plays an important role in the mitochondrial respiratory chain by transferring electrons from the Rieske iron-sulfur protein to cytochrome c. Mutations in this gene may cause mitochondrial complex III deficiency, nuclear type 6.