Learn More
ABclonal Technology JMJD8 Rabbit pAb
SDP
Catalog No. 5000137668
Encompass_Preferred
Involved in several processes, including positive regulation of I-kappaB kinase/NF-kappaB signaling, positive regulation of sprouting angiogenesis, and regulation of glycolytic process. Located in endoplasmic reticulum lumen and nucleus.
Catalog No. 50-001-37668 Supplier ABclonal Technology Supplier No. A104761000UL
Involved in several processes, including positive regulation of I-kappaB kinase/NF-kappaB signaling, positive regulation of sprouting angiogenesis, and regulation of glycolytic process. Located in endoplasmic reticulum lumen and nucleus.