Learn More
ABclonal Technology NOXO1 Rabbit pAb
SDP
Catalog No. 5000138092
Encompass_Preferred
This gene encodes an NADPH oxidase (NOX) organizer, which positively regulates NOX1 and NOX3. The protein contains a PX domain and two SH3 domains. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
Catalog No. 50-001-38092 Supplier ABclonal Technology Supplier No. A104771000UL
This gene encodes an NADPH oxidase (NOX) organizer, which positively regulates NOX1 and NOX3. The protein contains a PX domain and two SH3 domains. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.