Learn More
ABclonal Technology COPB1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A104851000UL
Encompass_Preferred
This gene encodes a protein subunit of the coatomer complex associated with non-clathrin coated vesicles. The coatomer complex, also known as the coat protein complex 1, forms in the cytoplasm and is recruited to the Golgi by activated guanosine triphosphatases. Once at the Golgi membrane, the coatomer complex may assist in the movement of protein and lipid components back to the endoplasmic reticulum. Alternatively spliced transcript variants have been described.
Catalog No. 50-001-36519