Learn More
ABclonal Technology DNAJC5 Rabbit pAb
SDP
Catalog No. 5000136517
Encompass_Preferred
This gene is a member of the J protein family. J proteins function in many cellular processes by regulating the ATPase activity of 70 kDa heat shock proteins. The encoded protein plays a role in membrane trafficking and protein folding, and has been shown to have anti-neurodegenerative properties. The encoded protein is known to play a role in cystic fibrosis and Huntingtons disease. A pseudogene of this gene is located on the short arm of chromosome 8.
Catalog No. 50-001-36517 Supplier ABclonal Technology Supplier No. A10489500UL
This gene is a member of the J protein family. J proteins function in many cellular processes by regulating the ATPase activity of 70 kDa heat shock proteins. The encoded protein plays a role in membrane trafficking and protein folding, and has been shown to have anti-neurodegenerative properties. The encoded protein is known to play a role in cystic fibrosis and Huntingtons disease. A pseudogene of this gene is located on the short arm of chromosome 8.