Learn More
ABclonal Technology MTAP Rabbit pAb
SDP
Catalog No. 5000138142
Encompass_Preferred
This gene encodes an enzyme that plays a major role in polyamine metabolism and is important for the salvage pathway of both adenine and methionine. The encoded enzyme is deficient in many cancers. Multiple alternatively spliced transcript variants have been described for this gene.
Catalog No. 50-001-38142 Supplier ABclonal Technology Supplier No. A10491000UL
This gene encodes an enzyme that plays a major role in polyamine metabolism and is important for the salvage pathway of both adenine and methionine. The encoded enzyme is deficient in many cancers. Multiple alternatively spliced transcript variants have been described for this gene.