Learn More
ABclonal Technology MTAP Rabbit pAb
SDP
Supplier:  ABclonal Technology A1049500UL
Encompass_Preferred
This gene encodes an enzyme that plays a major role in polyamine metabolism and is important for the salvage pathway of both adenine and methionine. The encoded enzyme is deficient in many cancers. Multiple alternatively spliced transcript variants have been described for this gene.
Catalog No. 50-001-37658