Learn More
ABclonal Technology ANO1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A10498500UL
Encompass_Preferred
Enables calcium activated cation channel activity, intracellular calcium activated chloride channel activity, and iodide transmembrane transporter activity. Involved in cation transport, inorganic anion transport, and positive regulation of insulin secretion involved in cellular response to glucose stimulus. Located in apical plasma membrane and nucleoplasm.
Catalog No. 50-001-36520