Learn More
ABclonal Technology CSF2RB Rabbit pAb
SDP
Supplier:  ABclonal Technology A105011000UL
Encompass_Preferred
The protein encoded by this gene is the common beta chain of the high affinity receptor for IL-3, IL-5 and CSF. Defects in this gene have been reported to be associated with protein alveolar proteinosis (PAP).
Catalog No. 50-001-35134