Learn More
ABclonal Technology CSF2RB Rabbit pAb
SDP
Catalog No. 5000135276
Encompass_Preferred
The protein encoded by this gene is the common beta chain of the high affinity receptor for IL-3, IL-5 and CSF. Defects in this gene have been reported to be associated with protein alveolar proteinosis (PAP).
Catalog No. 50-001-35276 Supplier ABclonal Technology Supplier No. A10501500UL
The protein encoded by this gene is the common beta chain of the high affinity receptor for IL-3, IL-5 and CSF. Defects in this gene have been reported to be associated with protein alveolar proteinosis (PAP).