Learn More
ABclonal Technology MPP6 Rabbit pAb
SDP
Supplier:  ABclonal Technology A10509500UL
Encompass_Preferred
Members of the peripheral membrane-associated guanylate kinase (MAGUK) family function in tumor suppression and receptor clustering by forming multiprotein complexes containing distinct sets of transmembrane, cytoskeletal, and cytoplasmic signaling proteins. All MAGUKs contain a PDZ-SH3-GUK core and are divided into 4 subfamilies, DLG-like (see DLG1, MIM 601014), ZO1-like (see TJP1, MIM 601009), p55-like (see MPP1, MIM 305360), and LIN2-like (see CASK, MIM 300172), based on their size and the presence of additional domains. MPP6 is a member of the p55-like MAGUK subfamily (Tseng et al., 2001 [PubMed 11311936]).
Catalog No. 50-001-38156