Learn More
ABclonal Technology GIPC1 Rabbit pAb
SDP
Catalog No. 5000137848
Encompass_Preferred
GIPC1 is a scaffolding protein that regulates cell surface receptor expression and trafficking (Lee et al., 2008 [PubMed 18775991]).
Catalog No. 50-001-37848 Supplier ABclonal Technology Supplier No. A10550500UL
GIPC1 is a scaffolding protein that regulates cell surface receptor expression and trafficking (Lee et al., 2008 [PubMed 18775991]).