Learn More
ABclonal Technology COPG1 Rabbit pAb
SDP
Catalog No. 5000138159
Encompass_Preferred
Predicted to enable structural molecule activity. Predicted to be involved in several processes, including Golgi vesicle transport, establishment of Golgi localization, and organelle transport along microtubule. Located in Golgi apparatus.
Catalog No. 50-001-38159 Supplier ABclonal Technology Supplier No. A10551500UL
Predicted to enable structural molecule activity. Predicted to be involved in several processes, including Golgi vesicle transport, establishment of Golgi localization, and organelle transport along microtubule. Located in Golgi apparatus.