Learn More
ABclonal Technology CREB3L2 Rabbit pAb
SDP
Supplier:  ABclonal Technology A105541000UL
Encompass_Preferred
This gene encodes a member of the oasis bZIP transcription factor family. Members of this family can dimerize but form homodimers only. The encoded protein is a transcriptional activator. Translocations between this gene on chromosome 7 and the gene fused in sarcoma on chromosome 16 can be found in some tumors. Multiple transcript variants encoding different isoforms have been found for this gene.
Catalog No. 50-001-35295