Learn More
ABclonal Technology BHLHE41 Rabbit pAb
SDP
Supplier:  ABclonal Technology A105551000UL
Encompass_Preferred
This gene encodes a basic helix-loop-helix protein expressed in various tissues. The encoded protein can interact with ARNTL or compete for E-box binding sites in the promoter of PER1 and repress CLOCK/ARNTLs transactivation of PER1. This gene is believed to be involved in the control of circadian rhythm and cell differentiation. Defects in this gene are associated with the short sleep phenotype.
Catalog No. 50-001-37692