Learn More
ABclonal Technology Slug Rabbit pAb
SDP
Supplier:  ABclonal Technology A10571000UL
Encompass_Preferred
This gene encodes a member of the Snail family of C2H2-type zinc finger transcription factors. The encoded protein acts as a transcriptional repressor that binds to E-box motifs and is also likely to repress E-cadherin transcription in breast carcinoma. This protein is involved in epithelial-mesenchymal transitions and has antiapoptotic activity. Mutations in this gene may be associated with sporatic cases of neural tube defects.
Catalog No. 50-001-36752