Learn More
ABclonal Technology SEC14L2 Rabbit pAb
SDP
Supplier:  ABclonal Technology A105861000UL
Encompass_Preferred
This gene encodes a cytosolic protein which belongs to a family of lipid-binding proteins including Sec14p, alpha-tocopherol transfer protein, and cellular retinol-binding protein. The encoded protein stimulates squalene monooxygenase which is a downstream enzyme in the cholesterol biosynthetic pathway. Alternatively spliced transcript variants encoding different isoforms have been identified for this gene.
Catalog No. 50-001-37686