Learn More
ABclonal Technology C5AR2 Rabbit pAb
SDP
Supplier:  ABclonal Technology A10588500UL
Encompass_Preferred
This gene encodes a G-protein coupled receptor 1 family member involved in the complement system of the innate immune response. Unlike classical G-protein coupled receptors, the encoded protein does not associate with intracellular G-proteins. It may instead modulate signal transduction through the beta-arrestin pathway, and may alternatively act as a decoy receptor. This gene may be involved in coronary artery disease and in the pathogenesis of sepsis. Alternative splicing results in multiple transcript variants.
Catalog No. 50-001-37886