Learn More
ABclonal Technology [KD Validated] MLST8 Rabbit pAb
SDP
Catalog No. 5000137550
Encompass_Preferred
Enables protein serine/threonine kinase activator activity. Involved in TORC1 signaling, positive regulation of TOR signaling, and regulation of actin cytoskeleton organization. Part of TORC1 complex and TORC2 complex.
Catalog No. 50-001-37550 Supplier ABclonal Technology Supplier No. A10591000UL
Enables protein serine/threonine kinase activator activity. Involved in TORC1 signaling, positive regulation of TOR signaling, and regulation of actin cytoskeleton organization. Part of TORC1 complex and TORC2 complex.